Anti-Salmonella typhimurium 0-4 Mouse Monoclonal Antibody [clone: 1E6]
Catalog # ABCAAB8274-250
Supplier: Abcam
Specifications
- Druh protilátky:Primární
- Symbol antigenu:Salmonella typhimurium 0-4
- Klonovalita:Monoclonal
- Klon:1E6
- Spojení:Unconjugated
- ELISA:Yes
- Průtoková cytometrie:Yes
- Hostitel:Mouse
- Imunochemie:Yes
- ImunoFluorescence:Yes
- Western blot:Yes
- Skladovací teplota:Cool
- Transportní teplota:Ice
- Rozměry protilátky:250 µg
- Bal.:250 µG
Specifications
About this item
Caution:
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Salmonella typhimurium 0-4
Clonality: monoclonal
Clone: 1000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: