Order Entry
Czech Republic
ContactUsLinkComponent
Anti-IgG Rabbit Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Anti-IgG Rabbit Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Catalog # ABCAAB6924-1
Supplier:  Abcam
Anti-IgG Rabbit Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Catalog # ABCAAB6924-1
Supplier:  Abcam

Specifications

  • Druh protilátky:
    Sekundární
  • Symbol antigenu:
    IgG
  • Klonovalita:
    Polyclonal
  • Spojení:
    TRITC (Tetramethylrhodamine Isothiocyanate)
  • ELISA:
    Yes
  • Průtoková cytometrie:
    Yes
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • ImunoFluorescence:
    Yes
  • Skladovací teplota:
    Cool
  • Transportní teplota:
    Ice
  • Rozměry protilátky:
    1 mg
  • Bal.:
    1 mg

Specifications

About this item

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Secondary
Antigen: IgG (Bovine) (bovine IgG)
Clonality: polyclonal
Clone:
Conjugation: TRITC (Rhodamine)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: