Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-IgG Donkey Polyclonal Antibody (Biotinylated)
  ABCAAB6899-1
 :  Abcam
undefined
Anti-IgG Donkey Polyclonal Antibody (Biotinylated)
  ABCAAB6899-1
 :  Abcam

 

  • Druh protilátky:
    Sekundární
  • Symbol antigenu:
    IgG
  • Klonovalita:
    Polyclonal
  • Spojení:
    Biotin
  • ELISA:
    Yes
  • Hostitel:
    Donkey
  • Imunochemie:
    Yes
  • ImunoFluorescence:
    Yes
  • Western blot:
    Yes
  • Skladovací teplota:
    Cool
  • Transportní teplota:
    Ice
  • Rozměry protilátky:
    1 mg
  • Bal.:
    1 mg

 

 

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Secondary
Antigen: IgG (Sheep) (sheep IgG)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Donkey
Isotype: none
Reactivity: