Anti-IgG Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Katalogové číslo ABCAAB6270-500
Dodavatel: Abcam
Specifikace
- Druh protilátky:Sekundární
- Symbol antigenu:IgG
- Klonovalita:Polyclonal
- Spojení:TRITC (Tetramethylrhodamine Isothiocyanate)
- ELISA:Yes
- Průtoková cytometrie:Yes
- Hostitel:Goat
- Imunochemie:Yes
- ImunoFluorescence:Yes
- Izotyp:IgG
- Skladovací teplota:Cool
- Transportní teplota:Ice
- Rozměry protilátky:500 µg
- Bal.:500 µG
Specifikace
O tomto produktu
Caution:
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Secondary
Antigen: IgG (Rat) (rat IgG)
Clonality: polyclonal
Clone:
Conjugation: TRITC (Rhodamine)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: