Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-MCM2 Goat Polyclonal Antibody
  ABCAAB3723-50
 :  Abcam
undefined
Anti-MCM2 Goat Polyclonal Antibody
  ABCAAB3723-50
 :  Abcam

 

  • Druh protilátky:
    Primární
  • Název antigenu:
    minichromosome maintenance complex component 2
  • Symbol antigenu:
    MCM2
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • Hostitel:
    Goat
  • Imunochemie:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human
  • Western blot:
    Yes
  • Skladovací teplota:
    Cool
  • Transportní teplota:
    Ice
  • Rozměry protilátky:
    50 µl
  • Bal.:
    50 µl

 

 

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: MCM2 (minichromosome maintenance complex component 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human