Order Entry
Czech Republic
ContactUsLinkComponent
Anti-alpha Adaptin Mouse Monoclonal Antibody [clone: AC1-M11]
Anti-alpha Adaptin Mouse Monoclonal Antibody [clone: AC1-M11]
Catalog # ABCAAB2807-500
Supplier:  Abcam
undefined
Anti-alpha Adaptin Mouse Monoclonal Antibody [clone: AC1-M11]
Catalog # ABCAAB2807-500
Supplier:  Abcam

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    alpha Adaptin
  • Klonovalita:
    Monoclonal
  • Klon:
    AC1-M11
  • Spojení:
    Unconjugated
  • Hostitel:
    Mouse
  • Imunochemie:
    Yes
  • ImunoFluorescence:
    Yes
  • Imunoprecipitace:
    Yes
  • Izotyp:
    IgG2a
  • Western blot:
    Yes
  • Skladovací teplota:
    Cool
  • Transportní teplota:
    Ice
  • Rozměry protilátky:
    100 µg
  • Typ balení:
    Vial
  • Bal.:
    500 µl

Specifications

About this item

Mouse monoclonal [AC1-M11] to alpha Adaptin.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: alpha Adaptin
Clonality: Monoclonal
Clone: AC1-M11
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat; Human; Mouse