Order Entry
Czech Republic
ContactUsLinkComponent
Anti-ITGA2B Mouse Monoclonal Antibody [clone: MEM-06]
Anti-ITGA2B Mouse Monoclonal Antibody [clone: MEM-06]
  ABCAAB2211-100
 :  Abcam
Anti-ITGA2B Mouse Monoclonal Antibody [clone: MEM-06]
  ABCAAB2211-100
 :  Abcam

 

  • Druh protilátky:
    Primární
  • Název antigenu:
    Integrin Alpha 2 Beta
  • Symbol antigenu:
    ITGA2B
  • Klonovalita:
    Monoclonal
  • Klon:
    MEM-06
  • Spojení:
    Unconjugated
  • Průtoková cytometrie:
    Yes
  • Hostitel:
    Mouse
  • Izotyp:
    IgG1
  • Reaktivita:
    Human
  • Skladovací teplota:
    Cool
  • Transportní teplota:
    Ice
  • Bal.:
    100 µG

 

 

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: ITGA2B (integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigen CD41))
Clonality: Monoclonal
Clone: MEM-06
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human