Anti-Serum Amyloid A Mouse Monoclonal Antibody [clone: 115]
Catalog # ABCAAB687-100
Supplier: Abcam
Specifications
- Druh protilátky:Primární
- Název antigenu:Serum Amyloid A1
- Symbol antigenu:SAA1
- Klonovalita:Monoclonal
- Klon:115
- Spojení:Unconjugated
- ELISA:Yes
- Hostitel:Mouse
- Imunochemie:Yes
- Izotyp:IgG1
- Reaktivita:Human
- Western blot:Yes
- Skladovací teplota:Cool
- Transportní teplota:Ice
- Rozměry protilátky:100 µg
- Typ balení:Vial
- Bal.:100 µG
Specifications
About this item
Mouse monoclonal [115] to Serum Amyloid A.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Serum Amyloid A
Clonality: Monoclonal
Clone: 115
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human