Anti-RAD51 Mouse Monoclonal Antibody [clone: 14B4]
Katalogové číslo ABCAAB213-100
Dodavatel: Abcam
Specifikace
- Druh protilátky:Primární
- Název antigenu:RAD51 recombinase
- Symbol antigenu:RAD51
- Klonovalita:Monoclonal
- Klon:14B4
- Spojení:Unconjugated
- Hostitel:Mouse
- Imunochemie:Yes
- ImunoFluorescence:Yes
- Imunoprecipitace:Yes
- Izotyp:IgG2b
- Reaktivita:Human
- Western blot:Yes
- Skladovací teplota:Cool
- Transportní teplota:Ice
- Rozměry protilátky:100 µg
- Bal.:100 µl
Specifikace
O tomto produktu
Caution:
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: RAD51 (RAD51 recombinase)
Clonality: monoclonal
Clone: 14B4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human