Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Synaptotagmin Rabbit Polyclonal Antibody
Anti-Synaptotagmin Rabbit Polyclonal Antibody
Catalog # BIRBORB99276-100
Supplier:  Biorbyt
undefined
Anti-Synaptotagmin Rabbit Polyclonal Antibody
Catalog # BIRBORB99276-100
Supplier:  Biorbyt

Specifications

  • Druh protilátky:
    Primární
  • Symbol antigenu:
    Synaptotagmin
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Modifikace:
    Phospho-Thr202
  • Skladovací pufr:
    phosphate buffer saline, pH. 7.4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
  • Molekulární hmotnost:
    60 kDa
  • Skladovací teplota:
    Cool
  • Koncentrace:
    20 ug: 0,2 mg/ml; 50 ug: 0,5 mg/ml; 100 ug: 1 ug/ul; 200 ug: 1 ug/ul
  • Transportní teplota:
    Ice
  • Imunogen:
    A synthesized peptide derived from human Synaptotagmin around the phosphorylation site of Threonine 202.
  • Čištění:
    purified by immunoaffinity chromatography
  • Rozměry protilátky:
    100 µg
  • Bal.:
    100 µG

Specifications

About this item

Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200

Type: Primary
Antigen: Synaptotagmin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat