Anti-MAPK1/3 Rabbit Polyclonal Antibody
Catalog # BIRBORB99245-100
Supplier: Biorbyt
Specifications
- Druh protilátky:Primární
- Název antigenu:Mitogen-activated protein kinase 1/3
- Symbol antigenu:MAPK1/3
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- Hostitel:Rabbit
- Imunochemie:Yes
- Izotyp:IgG
- Reaktivita:Human,Rat,Mouse
- Western blot:Yes
- Modifikace:Phospho-Thr202
- Skladovací pufr:phosphate buffer saline, pH. 7.4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Molekulární hmotnost:42, 44 kDa
- Skladovací teplota:Cool
- Koncentrace:20 ug: 0,2 mg/ml; 50 ug: 0,5 mg/ml; 100 ug: 1 ug/ul; 200 ug: 1 ug/ul
- Transportní teplota:Ice
- Imunogen:A synthesized peptide derived from human p44/42 MAP Kinase around the phosphorylation site of Thr202
- Čištění:purified by immunoaffinity chromatography
- Rozměry protilátky:100 µg
- Bal.:100 µG
Specifications
About this item
Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:200
Type: Primary
Antigen: MAPK1/3 (mitogen-activated protein kinase 1/3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat