Order Entry
Czech Republic
ContactUsLinkComponent
Anti-GATA4 Rabbit polyclonal antibody unconjugated
Anti-GATA4 Rabbit polyclonal antibody unconjugated
Catalog # BIRBORB99149-100
Supplier:  Biorbyt
undefined
Anti-GATA4 Rabbit polyclonal antibody unconjugated
Catalog # BIRBORB99149-100
Supplier:  Biorbyt

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    GATA binding Protein 4
  • Symbol antigenu:
    GATA4
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • Hostitel:
    Rabbit
  • ImunoFluorescence:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Modifikace:
    phospho-Ser262
  • Skladovací pufr:
    Phosphate buffer saline, pH. 7.4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
  • Molekulární hmotnost:
    48 kDa
  • Skladovací teplota:
    Cool
  • Koncentrace:
    20 ug: 0,2 mg/ml; 50 ug: 0,5 mg/ml; 100 ug: 1 ug/ul; 200 ug: 1 ug/ul
  • Transportní teplota:
    Ice
  • Imunogen:
    A synthesized peptide derived from human GATA4 around the phosphorylation site of Serine 262 .
  • Čištění:
    purified by immunoaffinity chromatography
  • Rozměry protilátky:
    100 µg
  • Bal.:
    100 µG

Specifications

About this item

Western Blot: 1:500-1:2000; Immunofluorescence: 1:100-1:500

Type: Primary
Antigen: GATA4 (GATA binding protein 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat