Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CSF1R Rabbit polyclonal antibody unconjugated
Anti-CSF1R Rabbit polyclonal antibody unconjugated
  BIRBORB97031-100
 :  Biorbyt
undefined
Anti-CSF1R Rabbit polyclonal antibody unconjugated
  BIRBORB97031-100
 :  Biorbyt

 

  • Druh protilátky:
    Primární
  • Název antigenu:
    colony stimulating factor 1 receptor
  • Symbol antigenu:
    CSF1R
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • Hostitel:
    Rabbit
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Formulář:
    lyophilized
  • Skladovací pufr:
    5mg BSA, 0,9mg NaCl, 0,2mg Na2HPO4, 0,05mg Thimerosal, 0,05mg NaN3.
  • Skladovací teplota:
    Cool
  • Koncentrace:
    0,1-0,5 ug/ml
  • Transportní teplota:
    Ice
  • Imunogen:
    A synthetic peptide corresponding to a sequence at the C-terminal of human CSF1R, different from the related mouse sequence by one amino acid and from the related rat sequence by two amino acids.
  • Čištění:
    purified by immunoaffinity chromatography
  • Rozměry protilátky:
    100 µg
  • Bal.:
    100 µG

 

 

Type: Primary
Antigen: CSF1R (colony stimulating factor 1 receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat