Order Entry
Czech Republic
ContactUsLinkComponent
Anti-STAT1 Rabbit Polyclonal Antibody
Anti-STAT1 Rabbit Polyclonal Antibody
Catalog # BIRBORB86788-50
Supplier:  Biorbyt
undefined
Anti-STAT1 Rabbit Polyclonal Antibody
Catalog # BIRBORB86788-50
Supplier:  Biorbyt

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    Signal Transducer And Activator of Transcription 1
  • Symbol antigenu:
    STAT1
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • ImunoFluorescence:
    Yes
  • Imunoprecipitace:
    Yes
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Skladovací pufr:
    PBS, 0,02% sodium azide.
  • Skladovací teplota:
    Cool
  • Transportní teplota:
    Ice
  • Čištění:
    purified by DEAE chromatography
  • Rozměry protilátky:
    50 µg
  • Bal.:
    50 µG

Specifications

About this item

Immunochemistry: 5 ?g/ml: 1:1000-1:2000

Type: Primary
Antigen: STAT1 (signal transducer and activator of transcription 1, 91kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat