Order Entry
Czech Republic
ContactUsLinkComponent
Anti-RABEPK Rabbit Polyclonal Antibody
Anti-RABEPK Rabbit Polyclonal Antibody
Catalog # BIRBORB78206-100
Supplier:  Biorbyt
undefined
Anti-RABEPK Rabbit Polyclonal Antibody
Catalog # BIRBORB78206-100
Supplier:  Biorbyt

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    Rab9 effector Protein with kelch motifs
  • Symbol antigenu:
    RABEPK
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • ELISA:
    Yes
  • Hostitel:
    Rabbit
  • Izotyp:
    IgG
  • Reaktivita:
    Human
  • Western blot:
    Yes
  • Skladovací teplota:
    Cool
  • Transportní teplota:
    Ice
  • Imunogen:
    Recombinant protein
  • Čištění:
    purified by affinity chromatography
  • Rozměry protilátky:
    100 µg
  • Bal.:
    100 µG

Specifications

About this item

ELISA: 1:20000-1:80000; Western Blot: 1:1000-1:2000

Type: Primary
Antigen: RABEPK (Rab9 effector protein with kelch motifs)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human