Anti-PITPNM2 Rabbit Polyclonal Antibody
Catalog # BIRBORB78135-100
Supplier: Biorbyt
Specifications
- Druh protilátky:Primární
- Název antigenu:phosphatidylinositol transfer Protein, membrane-associated 2
- Symbol antigenu:PITPNM2
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- ELISA:Yes
- Hostitel:Rabbit
- Imunochemie:Yes
- Izotyp:IgG
- Reaktivita:Human
- Western blot:Yes
- Skladovací teplota:Cool
- Transportní teplota:Ice
- Imunogen:Recombinant protein
- Čištění:purified by affinity chromatography
- Rozměry protilátky:100 µg
- Bal.:100 µG
Specifications
About this item
- ELISA: 1:20000-1:80000; Western Blot: 1:500-1:1000
Type: Primary
Antigen: PITPNM2 (phosphatidylinositol transfer protein, membrane-associated 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human