Anti-CLEC2A Rabbit Polyclonal Antibody
Catalog # BIRBORB75821-100
Supplier: Biorbyt
Specifications
- Druh protilátky:Primární
- Název antigenu:C-type lectin domain family 2, member A
- Symbol antigenu:CLEC2A
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- ELISA:Yes
- Hostitel:Rabbit
- Reaktivita:Human
- Western blot:Yes
- Skladovací teplota:Cool
- Koncentrace:1 mg/mL
- Transportní teplota:Ice
- Rozměry protilátky:100 µg
- Bal.:100 µG
Specifications
About this item
Type: Primary
Antigen: CLEC2A (C-type lectin domain family 2, member A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human