Anti-MMP2 Rabbit Polyclonal Antibody
Catalog # BIRBORB74163-100
Supplier: Biorbyt
Specifications
- Druh protilátky:Primární
- Název antigenu:matrix metallopeptidase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IV collagenase)
- Symbol antigenu:MMP2
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- Hostitel:Rabbit
- ImunoFluorescence:Yes
- Reaktivita:Human
- Western blot:Yes
- Skladovací teplota:Cool
- Koncentrace:0,55 mg/ml
- Transportní teplota:Ice
- Rozměry protilátky:100 µl
- Bal.:100 µl
Specifications
About this item
Type: Primary
Antigen: MMP2 (matrix metallopeptidase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IV collagenase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human