Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Bisphosphoglycerate mutase Rabbit Polyclonal Antibody
Anti-Bisphosphoglycerate mutase Rabbit Polyclonal Antibody
Catalog # BIRBORB74092-100
Supplier:  Biorbyt
undefined
Anti-Bisphosphoglycerate mutase Rabbit Polyclonal Antibody
Catalog # BIRBORB74092-100
Supplier:  Biorbyt

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    Bisphosphoglycerate mutase
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • Hostitel:
    Rabbit
  • ImunoFluorescence:
    Yes
  • Reaktivita:
    Human
  • Western blot:
    Yes
  • Skladovací teplota:
    Cool
  • Koncentrace:
    0,84 mg/ml
  • Transportní teplota:
    Ice
  • Rozměry protilátky:
    100 µl
  • Bal.:
    100 µl

Specifications

About this item

Type: Primary
Antigen: Bisphosphoglycerate mutase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human