Anti-WNT11 Rabbit Polyclonal Antibody
BIRBORB73750-100
: Biorbyt
- Druh protilátky:Primární
- Název antigenu:wingless-type MMTV integration site family, member 11
- Symbol antigenu:WNT11
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- Hostitel:Rabbit
- Imunochemie:Yes
- ImunoFluorescence:Yes
- Reaktivita:Human
- Western blot:Yes
- Skladovací teplota:Cool
- Koncentrace:1 mg/ml
- Transportní teplota:Ice
- Rozměry protilátky:100 µl
- Bal.:100 µl
Type: Primary
Antigen: WNT11 (wingless-type MMTV integration site family, member 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human