Anti-HSPA5 Rabbit Polyclonal Antibody
: Biorbyt
Western Blot: 1:1000-1:2000
Type: Primary
Antigen: HSPA5 (heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
- Catalog No:
- BIRBORB67549-100
- BIRBORB67549-25
- Symbol antigenu:
- HSPA5
- HSPA5
- Název antigenu:
- Heat shock 70kDa protein 5
- Heat shock 70kDa protein 5
- Druh protilátky:
- Primární
- Primární
- Klonovalita:
- Polyclonal
- Polyclonal
- Spojení:
- Unconjugated
- Unconjugated
- Hostitel:
- Rabbit
- Rabbit
- Izotyp:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Imunogen:
- Full length human GRP78 (Bip) his tagged at the N terminus
- Full length human GRP78 (Bip) his tagged at the N terminus
- Čištění:
- purified by affinity chromatography
- purified by affinity chromatography
- Skladovací pufr:
- PBS, 50% glycerol, 0,09% sodium azide
- PBS, 50% glycerol, 0,09% sodium azide
- Skladovací teplota:
- Cool
- Cool
- Transportní teplota:
- Ice
- Ice
Frequently Bought Together