Order Entry
Czech Republic
ContactUsLinkComponent
Anti-SMARCB1 Rabbit Polyclonal Antibody
Anti-SMARCB1 Rabbit Polyclonal Antibody
Catalog # BIRBORB67067-100
Supplier:  Biorbyt
undefined
Anti-SMARCB1 Rabbit Polyclonal Antibody
Catalog # BIRBORB67067-100
Supplier:  Biorbyt

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily b, member 1
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • Hostitel:
    Rabbit
  • Izotyp:
    IgG
  • Reaktivita:
    Human
  • Western blot:
    Yes
  • Skladovací pufr:
    1 x PBS (pH 7,4), 40% Glycerol, 0,02% Thimerosal.
  • Skladovací teplota:
    Cool
  • Transportní teplota:
    Ice
  • Imunogen:
    N-terminal region of human SMARCB1
  • Čištění:
    purified by affinity chromatography
  • Rozměry protilátky:
    100 µl
  • Bal.:
    100 µl

Specifications

About this item

Western Blot: 1:1000-1:5000

Type: Primary
Antigen: SMARCB1 (SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily b, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human