Anti-NANS Mouse Monoclonal Antibody
BIRBORB137436-50
: Biorbyt
- Druh protilátky:Primární
- Název antigenu:N-acetylneuraminic acid synthase
- Symbol antigenu:NANS
- Klonovalita:Monoclonal
- Spojení:Unconjugated
- ELISA:Yes
- Hostitel:Mouse
- Imunochemie:Yes
- Izotyp:IgG1 kappa
- Reaktivita:Human
- Western blot:Yes
- Skladovací pufr:PBS, pH 7,4, 0,1% sodium azide
- Skladovací teplota:Cool
- Koncentrace:1 mg/ml
- Transportní teplota:Ice
- Imunogen:NANS antibody was raised against recombinant human NANS (1-359aa) purified from E. coli.
- Čištění:purified by Protein G chromatography
- Rozměry protilátky:50 µl
- Bal.:50 µl
Immunochemistry: 10 ?g/ml; Western Blot: 1:1000
Type: Primary
Antigen: NANS (N-acetylneuraminic acid synthase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human