Order Entry
Czech Republic
ContactUsLinkComponent
Anti-SLCO2B1 Rabbit Polyclonal Antibody (RBITC)
Anti-SLCO2B1 Rabbit Polyclonal Antibody (RBITC)
Catalog # BIRBORB131615-100
Supplier:  Biorbyt
Anti-SLCO2B1 Rabbit Polyclonal Antibody (RBITC)
Catalog # BIRBORB131615-100
Supplier:  Biorbyt

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    solute carrier organic anion transporter family, member 2B1
  • Klonovalita:
    Polyclonal
  • Spojení:
    RBITC (Rhodamine B Isothiocyanate)
  • Hostitel:
    Rabbit
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Formulář:
    liquid
  • Skladovací pufr:
    10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • Molekulární hmotnost:
    77 kDa
  • Skladovací teplota:
    Cool
  • Transportní teplota:
    Ice
  • Imunogen:
    KLH conjugated synthetic peptide derived from human SLCO2B1.
  • Čištění:
    purified by affinity chromatography
  • Rozměry protilátky:
    100 µl
  • Bal.:
    100 µl

Specifications

About this item

Type: Primary
Antigen: SLCO2B1 (solute carrier organic anion transporter family, member 2B1)
Clonality: polyclonal
Clone:
Conjugation: RBITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat