Anti-KLRB1 Rabbit Polyclonal Antibody (RBITC)
Catalog # BIRBORB131264-100
Supplier: Biorbyt
Specifications
- Druh protilátky:Primární
- Název antigenu:killer cell lectin-like receptor subfamily B, member 1
- Symbol antigenu:KLRB1
- Klonovalita:Polyclonal
- Spojení:RBITC (Rhodamine B Isothiocyanate)
- Hostitel:Rabbit
- Izotyp:IgG
- Reaktivita:Human
- Formulář:liquid
- Skladovací pufr:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molekulární hmotnost:25 kDa
- Skladovací teplota:Cool
- Transportní teplota:Ice
- Imunogen:KLH conjugated synthetic peptide derived from hu CD161/NK1.
- Čištění:purified by affinity chromatography
- Rozměry protilátky:100 µl
- Bal.:100 µl
Specifications
About this item
Type: Primary
Antigen: KLRB1 (killer cell lectin-like receptor subfamily B, member 1)
Clonality: polyclonal
Clone:
Conjugation: RBITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human