Anti-EIF2AK4 Rabbit Polyclonal Antibody (PE (Phycoerythrin)/Cy5.5®)
BIRBORB130410-100
: Biorbyt
- Druh protilátky:Primární
- Název antigenu:Eukaryotic translation initiation factor 2 alpha kinase 4
- Symbol antigenu:EIF2AK4
- Klonovalita:Polyclonal
- Spojení:PE (Phycoerythrin)/Cy5.5®
- Hostitel:Rabbit
- Izotyp:IgG
- Reaktivita:Human,Rat,Mouse
- Formulář:liquid
- Modifikace:phospho-Thr899
- Skladovací pufr:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molekulární hmotnost:187 kDa
- Skladovací teplota:Cool
- Transportní teplota:Ice
- Imunogen:KLH conjugated synthetic phosphopeptide derived from human GCN2 around the phosphorylation site of Thr899.
- Čištění:purified by affinity chromatography
- Rozměry protilátky:100 µl
- Bal.:100 µl
Type: Primary
Antigen: EIF2AK4 (eukaryotic translation initiation factor 2 alpha kinase 4)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)/Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat