Anti-CDKN2A Rabbit Polyclonal Antibody
Supplier: Biorbyt
Certificates
About this item
Type: Primary
Antigen: CDKN2A (cyclin-dependent kinase inhibitor 2A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Specifications
- Catalog No:
- BIRBORB129709-100
- BIRBORB129709-200
- Symbol antigenu:
- CDKN2A
- CDKN2A
- Název antigenu:
- Cyclin-dependent kinase inhibitor 2A
- Cyclin-dependent kinase inhibitor 2A
- Druh protilátky:
- Primární
- Primární
- Klonovalita:
- Polyclonal
- Polyclonal
- Spojení:
- Unconjugated
- Unconjugated
- Hostitel:
- Rabbit
- Rabbit
- Western blot:
- Yes
- Yes
- Imunochemie:
- Yes
- Yes
- Imunogen:
- C term -peptide
- C term -peptide
- Čištění:
- purified by affinity chromatography
- purified by affinity chromatography
- Koncentrace:
- 0,8 mg/ml
- 0,8 mg/ml
- Skladovací teplota:
- Cool
- Cool
- Transportní teplota:
- Ice
- Ice
Specifications
Frequently Bought Together