Anti-HIST3H3 Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
Certifikáty
O tomto produktu
Type: Primary
Antigen: HIST3H3 (histone cluster 3, H3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Specifikace
- Catalog No:
- BIRBORB128179-200
- BIRBORB128179-100
- Symbol antigenu:
- HIST3H3
- HIST3H3
- Název antigenu:
- histone cluster 3, H3
- histone cluster 3, H3
- Druh protilátky:
- Primární
- Primární
- Klonovalita:
- Polyclonal
- Polyclonal
- Spojení:
- Unconjugated
- Unconjugated
- Reaktivita:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Hostitel:
- Rabbit
- Rabbit
- Western blot:
- Yes
- Yes
- Modifikace:
- phospho-Ser28
- phospho-Ser28
- Čištění:
- purified by affinity chromatography
- purified by affinity chromatography
- Koncentrace:
- 1 mg/ml
- 1 mg/ml
- Skladovací pufr:
- PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Skladovací teplota:
- Cool
- Cool
- Transportní teplota:
- Ice
- Ice
Specifikace
Frequently Bought Together