Anti-SPINK6 Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
Certifikáty
O tomto produktu
Type: Primary
Antigen: SPINK6 (serine peptidase inhibitor, Kazal type 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Specifikace
- Catalog No:
- BIRBORB126386-200
- BIRBORB126386-100
- Symbol antigenu:
- SPINK6
- SPINK6
- Název antigenu:
- Serine Peptidase Inhibitor, Kazal Type 6
- Serine Peptidase Inhibitor, Kazal Type 6
- Druh protilátky:
- Primární
- Primární
- Klonovalita:
- Polyclonal
- Polyclonal
- Spojení:
- Unconjugated
- Unconjugated
- Reaktivita:
- Human
- Human
- Hostitel:
- Rabbit
- Rabbit
- Imunochemie:
- Yes
- Yes
- Čištění:
- purified by affinity chromatography
- purified by affinity chromatography
- Koncentrace:
- 1 mg/ml
- 1 mg/ml
- Skladovací pufr:
- PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Skladovací teplota:
- Cool
- Cool
- Transportní teplota:
- Ice
- Ice