Anti-ITPKC Rabbit Polyclonal Antibody
Supplier: Biorbyt
Certificates
About this item
Type: Primary
Antigen: ITPKC (inositol-trisphosphate 3-kinase C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Specifications
- Catalog No:
- BIRBORB125899-100
- BIRBORB125899-200
- Symbol antigenu:
- ITPKC
- ITPKC
- Název antigenu:
- Inositol-trisphosphate 3-kinase C
- Inositol-trisphosphate 3-kinase C
- Druh protilátky:
- Primární
- Primární
- Klonovalita:
- Polyclonal
- Polyclonal
- Spojení:
- Unconjugated
- Unconjugated
- Reaktivita:
- Human
- Human
- Hostitel:
- Rabbit
- Rabbit
- Western blot:
- Yes
- Yes
- Čištění:
- purified by affinity chromatography
- purified by affinity chromatography
- Koncentrace:
- 1 mg/ml
- 1 mg/ml
- Skladovací pufr:
- PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Skladovací teplota:
- Cool
- Cool
- Transportní teplota:
- Ice
- Ice
Specifications
Frequently Bought Together