Anti-ATM Rabbit Polyclonal Antibody (Cy3®)
BIRBORB114458-100
: Biorbyt
- Druh protilátky:Primární
- Název antigenu:Ataxia Telangiectasia Mutated
- Symbol antigenu:ATM
- Klonovalita:Polyclonal
- Spojení:Cy3®
- Hostitel:Rabbit
- Izotyp:IgG
- Reaktivita:Human,Rat,Mouse
- Formulář:liquid
- Modifikace:phospho-Ser1981
- Skladovací pufr:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molekulární hmotnost:370 kDa
- Skladovací teplota:Cool
- Transportní teplota:Ice
- Imunogen:KLH conjugated synthetic phosphopeptide derived from human ATM around the phosphorylation site of Ser1981.
- Čištění:purified by affinity chromatography
- Rozměry protilátky:100 µl
- Bal.:100 µl
Type: Primary
Antigen: ATM (ataxia telangiectasia mutated)
Clonality: polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat