Anti-ALDH2 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Catalog # BIRBORB113451-100
Supplier: Biorbyt
Specifications
- Druh protilátky:Primární
- Název antigenu:aldehyde dehydrogenase 2 family (mitochondrial)
- Symbol antigenu:ALDH2
- Klonovalita:Polyclonal
- Spojení:AP (Alkaline Phosphatase)
- Hostitel:Rabbit
- Izotyp:IgG
- Reaktivita:Human,Rat,Mouse
- Formulář:liquid
- Skladovací pufr:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molekulární hmotnost:56 kDa
- Skladovací teplota:Cool
- Transportní teplota:Ice
- Imunogen:KLH conjugated synthetic peptide derived from human ALDHI.
- Čištění:purified by affinity chromatography
- Rozměry protilátky:100 µl
- Bal.:100 µl
Specifications
About this item
Type: Primary
Antigen: ALDH2 (aldehyde dehydrogenase 2 family (mitochondrial))
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat