Order Entry
Czech Republic
ContactUsLinkComponent
Anti-KCNMB1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-KCNMB1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BIRBORB110269-100
Supplier:  Biorbyt
Anti-KCNMB1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BIRBORB110269-100
Supplier:  Biorbyt

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    potassium large conductance calcium-activated channel, subfamily M, beta member 1
  • Symbol antigenu:
    KCNMB1
  • Klonovalita:
    Polyclonal
  • Spojení:
    HRP (Horseradish Peroxidase)
  • Hostitel:
    Rabbit
  • Izotyp:
    IgG
  • Formulář:
    liquid
  • Skladovací pufr:
    10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • Molekulární hmotnost:
    21 kDa
  • Skladovací teplota:
    Cool
  • Transportní teplota:
    Ice
  • Čištění:
    purified by affinity chromatography
  • Rozměry protilátky:
    100 µl
  • Bal.:
    100 µl

Specifications

About this item

Type: Primary
Antigen: KCNMB1 (potassium large conductance calcium-activated channel, subfamily M, beta member 1)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: