Anti-EIF4EBP1 Rabbit polyclonal antibody
100 µl
BIRBORB107133-100
Marketsource
: Biorbyt
Type: Primary
Antigen: EIF4EBP1 (eukaryotic translation initiation factor 4E binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
- Rozměry protilátky:100 µl
- Western blot:Yes
- Hostitel:Rabbit
- Klonovalita:Polyclonal
- Imunochemie:Yes
- Druh protilátky:Primární
- Modifikace:phospho-Thr36
- Symbol antigenu:EIF4EBP1
- Izotyp:IgG
- Čištění:purified by affinity chromatography
- Cat. No.:BIRBORB107133-100
- Reaktivita:Human|Rat|Mouse
- Transportní teplota:Ice
- Koncentrace:1 mg/ml
- Skladovací teplota:Cool
- Spojení:Unconjugated
- Název antigenu:Eukaryotic translation initiation factor 4E binding protein 1



