Anti-SART3 Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
Certifikáty
O tomto produktu
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunoprecipitation: 1:20-100; Immunochemistry: 1:100-500; IC 1:100-500; Immunofluorescence: 1:50-200
Type: Primary
Antigen: SART3 (squamous cell carcinoma antigen recognized by T cells 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Specifikace
- Catalog No:
- BIRBORB101918-200
- BIRBORB101918-100
- Symbol antigenu:
- SART3
- SART3
- Název antigenu:
- Squamous cell carcinoma antigen recognized by T cells 3
- Squamous cell carcinoma antigen recognized by T cells 3
- Druh protilátky:
- Primární
- Primární
- Klonovalita:
- Polyclonal
- Polyclonal
- Spojení:
- Unconjugated
- Unconjugated
- Hostitel:
- Rabbit
- Rabbit
- Izotyp:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Imunochemie:
- Yes
- Yes
- Formulář:
- liquid
- liquid
- Molekulární hmotnost:
- 110 kDa
- 110 kDa
- Čištění:
- purified by affinity chromatography
- purified by affinity chromatography
- Skladovací pufr:
- 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Skladovací teplota:
- Cool
- Cool
- Transportní teplota:
- Ice
- Ice
Specifikace
Frequently Bought Together