Anti-FLOT1 Rabbit polyclonal antibody
Dodavatel: Biorbyt
Certifikáty
O tomto produktu
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)
Type: Primary
Antigen: FLOT1 (flotillin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifikace
- Catalog No:
- BIRBORB101600-200
- BIRBORB101600-100
- Symbol antigenu:
- FLOT1
- FLOT1
- Název antigenu:
- Flotillin 1
- Flotillin 1
- Druh protilátky:
- Primární
- Primární
- Klonovalita:
- Polyclonal
- Polyclonal
- Spojení:
- Unconjugated
- Unconjugated
- Reaktivita:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Hostitel:
- Rabbit
- Rabbit
- Izotyp:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- Imunochemie:
- Yes
- Yes
- Formulář:
- liquid
- liquid
- Molekulární hmotnost:
- 47 kDa
- 47 kDa
- Čištění:
- purified by affinity chromatography
- purified by affinity chromatography
- Koncentrace:
- 0,5 mg/ml
- 0,5 mg/ml
- Skladovací pufr:
- 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Skladovací teplota:
- Cool
- Cool
- Transportní teplota:
- Ice
- Ice
Specifikace
Frequently Bought Together