Anti-ODC1 Rabbit Polyclonal Antibody
Catalog # BIRBORB40843-100
Supplier: Biorbyt
Specifications
- Druh protilátky:Primární
- Název antigenu:ornithine decarboxylase 1
- Symbol antigenu:ODC1
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- Hostitel:Rabbit
- Reaktivita:Human
- Western blot:Yes
- Počet aminokyselin:180 to 433
- Skladovací pufr:0,1M Tris, 0,1M Glycine, 10% Glycerol (pH7).
- Skladovací teplota:Cool
- Koncentrace:0,66 mg/ml
- Transportní teplota:Ice
- Imunogen:Recombinant protein fragment contain a sequence corresponding to a region within amino acids 180 and 433 of human ODC1.
- Čištění:purified by antigen affinity chromatography
- Rozměry protilátky:100 µl
- Bal.:100 µl
Specifications
About this item
Western Blot: 1:500-1:3000
Type: Primary
Antigen: ODC1 (ornithine decarboxylase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human