Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Scramblase 1 Mouse Monoclonal Antibody
Anti-Scramblase 1 Mouse Monoclonal Antibody
Catalog # BIRBORB24811-100
Supplier:  Biorbyt
undefined
Anti-Scramblase 1 Mouse Monoclonal Antibody
Catalog # BIRBORB24811-100
Supplier:  Biorbyt

Specifications

  • Druh protilátky:
    Primární
  • Symbol antigenu:
    Scramblase 1
  • Klonovalita:
    Monoclonal
  • Spojení:
    Unconjugated
  • ELISA:
    Yes
  • Hostitel:
    Mouse
  • Izotyp:
    IgG1
  • Reaktivita:
    Human
  • Western blot:
    Yes
  • Formulář:
    lyophilized
  • Skladovací pufr:
    2X PBS, PEG and Sucrose
  • Skladovací teplota:
    Cool
  • Koncentrace:
    100 ug/ml (prior to lyophilization)
  • Transportní teplota:
    Ice
  • Imunogen:
    Peptide conjugated to KLH.
  • Čištění:
    purified by Thiophilic adsorption and size exclusion chromatography
  • Rozměry protilátky:
    100 µg
  • Bal.:
    100 µG

Specifications

About this item

Western Blot: 1 ?g/ml

Type: Primary
Antigen: Scramblase 1
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human