Anti-C. trachomatis EB Goat Polyclonal Antibody
Catalog # BIRBORB23154-1
Supplier: Biorbyt
Specifications
- Druh protilátky:Primární
- Název antigenu:C. trachomatis EB
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- ELISA:Yes
- Hostitel:Goat
- ImunoFluorescence:Yes
- Skladovací pufr:0,01M PBS, pH 7,2This product contains no stabilizing proteins
- Skladovací teplota:Cool
- Koncentrace:4-5 mg/ml (OD280, E(0,1%)=1,4)
- Transportní teplota:Ice
- Imunogen:L2 and other serovar groups
- Čištění:purified by DEAE chromatography
- Rozměry protilátky:1 ml
- Bal.:1 mL
Specifications
About this item
Type: Primary
Antigen: C. trachomatis EB
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: