Anti-C. trachomatis EB Goat Polyclonal Antibody (FITC (Fluorescein))
Catalog # BIRBORB23150-1
Supplier: Biorbyt
Specifications
- Druh protilátky:Primární
- Název antigenu:C. trachomatis EB
- Klonovalita:Polyclonal
- Spojení:FITC (Fluorescein Isothiocyanate)
- Hostitel:Goat
- ImunoFluorescence:Yes
- Skladovací pufr:0,01M PBS, pH 7,2 containing 10 mg/ml BSA
- Skladovací teplota:Cool
- Koncentrace:4-5 mg/ml (OD280, E(0,1%)=1,4)
- Transportní teplota:Ice
- Imunogen:L2 and other serovar groups
- Rozměry protilátky:1 ml
- Bal.:1 mL
Specifications
About this item
Type: Primary
Antigen: C. trachomatis EB
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Goat
Isotype: none
Reactivity: