Anti-IgG3 Rabbit Polyclonal Antibody
Catalog # BIRBORB21360-10
Supplier: Biorbyt
Specifications
- Druh protilátky:Sekundární
- Symbol antigenu:IgG3
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- Hostitel:Rabbit
- Formulář:lyophilized
- Skladovací teplota:Cool
- Koncentrace:1 mg/ml, no foreign proteins added,
- Transportní teplota:Ice
- Imunogen:Pools of purified homogenous IgG3 isolated from pooled mouse serum. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Rozměry protilátky:10 mg
- Bal.:10 mg
Specifications
About this item
Type: Secondary
Antigen: IgG3 (Mouse) (mouse IgG3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: