Anti-FH Rabbit Polyclonal Antibody (Biotin)
Catalog # BIRBORB21162-1
Supplier: Biorbyt
Specifications
- Druh protilátky:Primární
- Název antigenu:Fumarate hydratase
- Symbol antigenu:FH
- Klonovalita:Polyclonal
- Spojení:Biotin
- Hostitel:Rabbit
- Reaktivita:Pig
- Formát:immunoconjugate
- Skladovací pufr:phosphate buffered saline
- Skladovací teplota:Cool
- Koncentrace:1 mg/ml, Biotin/IgG protein molar ratio (B/P) approximately 7,6, no foreign proteins added,
- Transportní teplota:Ice
- Imunogen:Fumarase isolated and purified from porcine heart. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Rozměry protilátky:1 ml
- Bal.:1 mL
Specifications
About this item
Type: Primary
Antigen: FH (fumarate hydratase)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Pig