Anti-L-Lactic Dehydrogenase Rabbit Polyclonal Antibody (Biotin)
Catalog # BIRBORB20910-1
Supplier: Biorbyt
Specifications
- Druh protilátky:Primární
- Název antigenu:L-Lactic Dehydrogenase
- Symbol antigenu:L-Lactic Dehydrogenase
- Klonovalita:Polyclonal
- Spojení:Biotin
- Hostitel:Rabbit
- Reaktivita:Bovine
- Formát:immunoconjugate
- Skladovací pufr:phosphate buffered saline
- Skladovací teplota:Cool
- Koncentrace:1 mg/ml, Biotin/IgG protein molar ratio (B/P) approximately 5,6, no foreign proteins added,
- Transportní teplota:Ice
- Imunogen:L-Lactic dehydrogenase isolated and purified from bovine heart. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Rozměry protilátky:1 ml
- Bal.:1 mL
Specifications
About this item
Type: Primary
Antigen: L-Lactic Dehydrogenase
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Bovine