Order Entry
Czech Republic
ContactUsLinkComponent
Anti-SLC50A1 Goat Polyclonal Antibody
Anti-SLC50A1 Goat Polyclonal Antibody
Catalog # BIRBORB18557-100
Supplier:  Biorbyt
Anti-SLC50A1 Goat Polyclonal Antibody
Catalog # BIRBORB18557-100
Supplier:  Biorbyt

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    solute carrier family 50 (sugar efflux transporter), member 1
  • Symbol antigenu:
    SLC50A1
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • ELISA:
    Yes
  • Hostitel:
    Goat
  • Reaktivita:
    Human
  • Skladovací pufr:
    Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA._x000D_
  • Skladovací teplota:
    Cool
  • Transportní teplota:
    Ice
  • Imunogen:
    Synthetic peptide (C Terminus)
  • Čištění:
    purified by ammonium sulphate precipitation followed by antigen affinity chromatography
  • Rozměry protilátky:
    100 µg
  • Bal.:
    100 µG

Specifications

About this item

Type: Primary
Antigen: SLC50A1 (solute carrier family 50 (sugar efflux transporter), member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human