Order Entry
Czech Republic
ContactUsLinkComponent
Anti-HCN4 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anti-HCN4 Rabbit Polyclonal Antibody (FITC (Fluorescein))
  BIRBORB15736-100
 :  Biorbyt
Anti-HCN4 Rabbit Polyclonal Antibody (FITC (Fluorescein))
  BIRBORB15736-100
 :  Biorbyt

 

  • Druh protilátky:
    Primární
  • Název antigenu:
    hyperpolarization activated cyclic nucleotide-gated potassium channel 4
  • Symbol antigenu:
    HCN4
  • Klonovalita:
    Polyclonal
  • Spojení:
    FITC (Fluorescein Isothiocyanate)
  • Hostitel:
    Rabbit
  • ImunoFluorescence:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Formulář:
    liquid
  • Skladovací pufr:
    10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • Molekulární hmotnost:
    130 kDa
  • Skladovací teplota:
    Cool
  • Koncentrace:
    0,5 mg/ml
  • Transportní teplota:
    Ice
  • Imunogen:
    Synthetic peptide
  • Čištění:
    purified by affinity chromatography
  • Rozměry protilátky:
    100 µg
  • Bal.:
    100 µG

 

 

Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: HCN4 (hyperpolarization activated cyclic nucleotide-gated potassium channel 4)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat