Anti-IGF1R Rabbit Polyclonal Antibody
Katalogové číslo BIRBORB14679-100
Dodavatel: Biorbyt
Specifikace
- Druh protilátky:Primární
- Název antigenu:Insulin-Like Growth Factor 1 Receptor
- Symbol antigenu:IGF1R
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- Hostitel:Rabbit
- Imunochemie:Yes
- Izotyp:IgG
- Reaktivita:Human,Rat,Mouse
- Western blot:Yes
- Skladovací pufr:PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Skladovací teplota:Cool
- Koncentrace:1 mg/ml
- Transportní teplota:Ice
- Imunogen:The antiserum was produced against synthesized non-phosphopeptide derived from human IGF1 Receptor around the phosphorylation site of tyrosine 1280 ( S -F-YP-Y-S ).
- Čištění:purified by affinity chromatography
- Rozměry protilátky:100 µg
- Bal.:100 µG
Specifikace
O tomto produktu
Western Blot: 1:500-1:1000 Immunochemistry: 1:50-1:100
Type: Primary
Antigen: IGF1R (insulin-like growth factor 1 receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat