Order Entry
Czech Republic
ContactUsLinkComponent
Anti-MAPK9 Rabbit Polyclonal Antibody
Anti-MAPK9 Rabbit Polyclonal Antibody
  BIRBORB14628-100
 :  Biorbyt
undefined
Anti-MAPK9 Rabbit Polyclonal Antibody
  BIRBORB14628-100
 :  Biorbyt

 

  • Druh protilátky:
    Primární
  • Název antigenu:
    Mitogen-activated protein kinase 9
  • Symbol antigenu:
    MAPK9
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Skladovací pufr:
    PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
  • Skladovací teplota:
    Cool
  • Koncentrace:
    1 mg/ml
  • Transportní teplota:
    Ice
  • Imunogen:
    The antiserum was produced against synthesized non-phosphopeptide derived from human SAPK/JNK around the phosphorylation site of threonine 183 (M -M-TP-P-Y).
  • Čištění:
    purified by affinity chromatography
  • Rozměry protilátky:
    100 µg
  • Bal.:
    100 µG

 

 

Western Blot: 1:500-1:1000 Immunochemistry: 1:50-1:100

Type: Primary
Antigen: MAPK9 (mitogen-activated protein kinase 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat