Anti-MYSM1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Catalog # BIRBORB14158-100
Supplier: Biorbyt
Specifications
- Druh protilátky:Primární
- Název antigenu:Myb-like, SWIRM and MPN domains 1
- Symbol antigenu:MYSM1
- Klonovalita:Polyclonal
- Spojení:FITC (Fluorescein Isothiocyanate)
- Hostitel:Rabbit
- ImunoFluorescence:Yes
- Izotyp:IgG
- Reaktivita:Human,Rat,Mouse
- Formulář:liquid
- Skladovací pufr:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molekulární hmotnost:91 kDa
- Skladovací teplota:Cool
- Koncentrace:0,5 mg/ml
- Transportní teplota:Ice
- Imunogen:Synthetic peptide
- Čištění:purified by affinity chromatography
- Rozměry protilátky:100 µg
- Bal.:100 µG
Specifications
About this item
Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)
Type: Primary
Antigen: MYSM1 (Myb-like, SWIRM and MPN domains 1)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat