Anti-PRKCA Rabbit Polyclonal Antibody
Catalog # BIRBORB6755-100
Supplier: Biorbyt
Specifications
- Druh protilátky:Primární
- Název antigenu:Protein Kinase C Alpha
- Symbol antigenu:PRKCA
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- Hostitel:Rabbit
- Imunochemie:Yes
- Izotyp:IgG
- Western blot:Yes
- Formulář:liquid
- Modifikace:phospho-Tyr658
- Skladovací pufr:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molekulární hmotnost:74 kDa
- Skladovací teplota:Cool
- Koncentrace:0,5 mg/ml
- Transportní teplota:Ice
- Imunogen:KLH conjugated synthetic phosphopeptide derived from human PRKCA around the phosphorylation site of Ser657+Tyr658.
- Čištění:purified by affinity chromatography
- Rozměry protilátky:100 µg
- Bal.:100 µG
Specifications
About this item
- ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: PRKCA (protein kinase C, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: