Anti-C20orf112 Rabbit Polyclonal Antibody
Supplier: Biorbyt
Certificates
About this item
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)
Type: Primary
Antigen: C20orf112 (chromosome 20 open reading frame 112)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Specifications
- Catalog No:
- BIRBORB2006-100
- BIRBORB2006-200
- Symbol antigenu:
- C20orf112
- C20orf112
- Název antigenu:
- Chromosome 20 open reading frame 112
- Chromosome 20 open reading frame 112
- Druh protilátky:
- Primární
- Primární
- Klonovalita:
- Polyclonal
- Polyclonal
- Spojení:
- Unconjugated
- Unconjugated
- Hostitel:
- Rabbit
- Rabbit
- Izotyp:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- Imunochemie:
- Yes
- Yes
- Imunogen:
- Synthetic peptide
- Synthetic peptide
- Formulář:
- liquid
- liquid
- Čištění:
- purified by Protein A chromatography
- purified by Protein A chromatography
- Koncentrace:
- 0,5 mg/ml
- 0,5 mg/ml
- Skladovací pufr:
- 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Skladovací teplota:
- Cool
- Cool
- Transportní teplota:
- Ice
- Ice
Specifications
Frequently Bought Together